Supelco® Organic Reference Standard, 1-Nonene

Leverandør: Merck

74323-5ML
74323-5MLEA 3070 NOK
74323-5ML
Supelco® Organic Reference Standard, 1-Nonene
Standarder Organiske standarder

  • GHS signal: GHS02, GHS08, GHS07


NB: P210 - P301 + P310 + P331 - P302 + P352 - P305 + P351 + P338

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f6045c116200310696d8a78429526a5d71cd04e&vid=6f6045c116200310696d8a78429526a5d71cd04e&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

M-Clarity quality level MQ100
Environmentally Preferable
Tetthet 0,73 g/ml at 25 °C (lit.)
CAS-nr 124-11-8
MDL No. MFCD00009568
Formel CH₃(CH₂)₆CH=CH₂
Molekylvekt 126,24
Smeltepunkt –81 °C (lit.)
kokepunkt 146 °C (lit.)
Hardhetsklasse beskrivelse H226 - H304 - H315 - H319 - H335
Flammepunkt 24 °C - Closed cup
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR