Organic reference standard, cis-3-Hexenyl acetate, analytical standard, Supelco®

Leverandør: Merck

74597-1ML
74597-1MLEA 735 NOK
74597-1ML
Organic reference standard, cis-3-Hexenyl acetate, analytical standard, Supelco®
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39386fc2208b0d665bbab2cafab8c5d94b74ddcf&vid=39386fc2208b0d665bbab2cafab8c5d94b74ddcf&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse cis-3-Hexenyl acetate
CAS-nr 3681-71-8
Formel C₈H₁₄O₂  
kokepunkt 75-76 °C/23 mm Hg(lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR