1-Octanesulphonic acid sodium salt monohydrate ≥99.0% (by titrimetric analysis) for ioneparkromatografi, Supelco®

Leverandør: Merck

74882-10G-F 74882-50G-F 74882-250G-F
74882-10G-FEA 3175 NOK
74882-10G-F 74882-50G-F 74882-250G-F
1-Octanesulphonic acid sodium salt monohydrate ≥99.0% (by titrimetric analysis) for ioneparkromatografi, Supelco®
1-Octanesulphonic acid sodium salt monohydrate
Formel: C₈H₁₇NaO₃S·1H₂O
MDL nummer: MFCD00007544
CAS-nummer: 207596-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dea818c4c50e33f0b0142cf2017a315735797dd5&vid=dea818c4c50e33f0b0142cf2017a315735797dd5&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR