Sodium octyl sulphate, Supelco®

Leverandør: Merck

Synonymer: Octyl sulphate, sodium salt, Octyl sulphuric acid salt

M-Clarity™ quality level = MQ100

75073-10G
75073-10GEA 4860 NOK
75073-10G
Sodium octyl sulphate, Supelco®
Sodium octyl sulphate
Formel: C₈H₁₇NaO₄S
MW: 232.28 g/mol
Smeltepunkt: 181…183 °C
Tetthet: 1.266 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007470
CAS-nummer: 142-31-4
EINECS: 205-535-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11075d1926e9fac49cbd10f57b3a106ab0981011&vid=2e55626d4de228367fc83a9de6747d853170b664&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR