4-Aminophenylphosphate sodium salt, hydrate ≥98.0% (by TLC) for analysis, electrochemical grade, Supelco®

Leverandør: Merck

Synonymer: 4-APP

75113-100MG
75113-100MGEA 1950 NOK
75113-100MG
4-Aminophenylphosphate sodium salt, hydrate ≥98.0% (by TLC) for analysis, electrochemical grade, Supelco®
4-Aminophenylphosphate sodium salt

4-Aminophenyl phosphate monosodium salt hydrate may be used as a reagent for the sensitive detection and estimation of tumor necrosis factor-alpha (TNF-α) in undiluted serum developed via the process of electrochemical enzyme-linked immunosorbent assay (ELISA).

Formel: C₆H₇NO₄P·Na·4.0H₂O
MW: 283.2 g/mol
CAS-nummer: 108084-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47d22e64e0ebb00247e57a4a2e41be9366543da3&vid=9af7cb566012997942bbfc6ee2de846ccde4bf8b&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR