Destillasjonskolber, DURAN®
Leverandør: DWK Life Sciences
216532807 216532901
752-0500EA 785 NOK
752-0500 214-0435
Destillasjonskolber, DURAN®
Kolber Destillasjonskolber
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.