Kongorødt, Supelco®

Leverandør: Merck
Danger

Synonymer: CI 22120

M-Clarity™ quality level = MQ100

75768-25MG
75768-25MGEA 1625 NOK
75768-25MG
Kongorødt, Supelco®
Kongorødt
Formel: C₃₂H₂₂N₆Na₂O₆S₂
Smeltepunkt: > 360 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004028
CAS-nummer: 573-58-0
EINECS: 209-358-4
Merck Indeks: 12,02562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b2e073f7e134e244e449ac0f8071774ba7da937&vid=ec4b5a585676a7e6953b9f429692e72504f26d5f&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR