EDTA dinatrium salt 0.01 mol/l (0.02 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

7613.1000EA 675 NOK
7613.1000
EDTA dinatrium salt 0.01 mol/l (0.02 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
EDTA dinatrium salt
Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3
Merck Indeks: 12,03556

Bestill nå

Spesifikasjon Testresultater

Molarity 0.0099 - 0.0101


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR