Experimentation kits, Quick PCR
Leverandør: EDVOTEK
372
763-0002EA 1700 NOK
763-0002
Experimentation kits, Quick PCR
Lærings materiell Læremateriale for biologi PCR Learning Activities
This experiment uses PCR to amplify a small section of Lambda DNA via a 2-step process, saving valuable classroom time and allowing completion of the lab in one session.
  • For 10 lab groups
  • Complete in 60 minutes
Also required: 5 to 50 µl adjustable micropipettes, tips, thermal cycler, electrophoresis tank, and power supply.
Leveringsinformasjon: Kit includes instructions, PCR beads, DNA template and primers, DNA size ladder, ultrapure water, wax beads, gel loading dye, agarose, electrophoresis buffer, and gel stain.