Ready-to-Load™ kits, the case of the invisible bands
Leverandør: EDVOTEK
S-52
763-0009EA 0 NOK
763-0009
Ready-to-Load™ kits, the case of the invisible bands
Lærings materiell Læremateriale for biologi Genetics Learning Activities
Experiment using fluorescence and electrophoresis to solve the mystery of the invisible bands.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank, power supply and long wave UV lamp.
Leveringsinformasjon: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.