Leaf structure model
Leverandør: 3B Scientific
1002504
763-0137EA 5450 NOK
763-0137
Leaf structure model
Modeller Botanikk
Fagus silvatica. Three dimensional model of an beech tree leaf at 1500X magnification.
Shows different cell types and stomata.