Blood typing kit and cards, Eldoncard
Leverandør: ELDON CARD
610-24
763-0279EA 1900 NOK
763-0279
Blood typing kit and cards, Eldoncard
Diagnostiske systemer Clinical Reagents and Kits
The presence or absence of agglutinates will show the blood type. The blood typing kit contains everything to type one person: 1 Eldoncard, 4 Eldonsticks, 1 alcohol swab, 2 safety lancets, 1 result.card. 2 micropipettes are supplied for 50 blood typings. Shelf-life 24 months at 5-37 °C. The foil is used to protect the cards after use.
  • Portable and compact
  • Blood grouping on location
  • Result after just 2 minutes
  • Easy to learn, easy to use
  • No need for refrigeration and daily controls
  • Whole blood can be used down to 5 μl
  • No need for separation of red cells and blood plasma