Gas sampling pump and tubes
Leverandør: GASTEC
GV-50PS
763-0292EA 5875 NOK
763-0292
Gas sampling pump and tubes
Gassdeteksjonssystemer
Gas detector tube system accurately measures vapour concentration levels and slight changes in concentration level.
These detector tubes are used in experiments like respiration, photosynthesis and combustion to determine concentration level.
Bestillingsinformasjon: Includes a sampling pump, detector tubes and tips for detector tubes. More detector tubes are available separately.