Model experiments, genetic experiments
Leverandør: GR NNA PLASTEMBALLAGE AB
191050-100
763-0295EA 0 NOK
763-0295
Model experiments, genetic experiments
Lærings materiell Læremateriale for biologi Genetics Learning Activities
Model experiments with practical examples of working methods in genetics. During the experiments, the students quickly make so many observations that their calculations of outcomes can be considered statistically reliable. The experimental kit contains four cans of tubes and colored beads for experiments in some of the different areas of genetics. Blue or brown eyes? Boy or girl? How many peas turn yellow? Trial description included.