Lens with variable focal length
Leverandør: IMBE HANDELSBOGALET
194570-10
763-0325EA 0 NOK
763-0325
Lens with variable focal length
Lærings materiell Læremateriale for fysikk Optics Learning Activities
Lens made of silicone rubber, mounted in a screen. The shape can be varied steplessly from convex to concave by changing the water volume in the lens. If an object is placed in front of the lens, the object can be projected on a posterior screen (retina). The focus is adjusted with the volume of water in the lens.