Avantor® Seradigm South America Origin Fetal Bovine Serum (FBS)
Leverandør: VWR Seradigm
76314-518EA 4000 NOK
76314-518 76314-514
Avantor® Seradigm South America Origin Fetal Bovine Serum (FBS)
Serum
Avantor® Seradigm South America origin FBS preserves more of the naturally-occurring growth factors that cells require to thrive, resulting in the highest possible quality sera for a price that fits your budget.
  • Industry-leading collection techniques at the source
  • Manufacturing techniques reduce bioburden and ensure homogeneity for every bottle and for every lot
  • Risk mitigation through a robust quality system
  • Proven track record for performance and consistency
  • Sourced from countries with health status classified as "Negligible BSE risk" by the World Health