Maltese cross tube
Leverandør: 3B Scientific
1000011
765-0089EA 15700 NOK
765-0089
Maltese cross tube
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
Evacuated glass flask containing i.a. electron gun with directly heated tungsten wire cathode and a cylindrical anode. The electron gun emits a divergent electron beam, aimed at an Maltese cross of aluminum placed in the middle of the tube. The cross can be connected electrically. At the same time, light from the filament is also directed towards the cross, so that a comparison can be made between the shadows of different types that appear on the luminescent surface at the far end of the tube. Specifications.
  • Unit: 150 × 100 × 60 mm
  • Laser diode: 13 × 40 mm
The following phenomenaMore Product Information