Electrical energy sources
Leverandør: CORNELSEN EXPERIMENTA
23000
765-0109EA 0 NOK
765-0109
Electrical energy sources
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
The kit contains about 15 different components with which a number of experiments can be performed. The components are designed so that students can quickly and easily perform their laboratory work. The kit can show how the following forms of energy are generated:
  • Thermal energy (thermocouple)
  • Radiant energy (solar cell)
  • Chemical energy (galvanic element)
  • Mechanical energy (induction)
To demonstrate that electrical energy is generated, the units can be connected to an included micromotor with propeller. Detailed instructions for use included. The kitMore Product Information