Insulated rod
Leverandør: CORNELSEN EXPERIMENTA
40598
765-0112EA 0 NOK
765-0112
Insulated rod
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
Plastic rod with metal tube equipped with an axial and a radial 4 mm connection.
  • Dimensions: 300 × 10 mm