Hydrostatic paradox
Leverandør: CORNELSEN EXPERIMENTA
44210
765-0138EA 0 NOK
765-0138
Hydrostatic paradox
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Shows that the bottom pressure in a liquid depends only on the height and not the shape of the container. The fluid pressure is transferred via a rubber diaphragm to an adjustable hand. Comes with four containers of different shapes. Dimensions.
  • Height: 290 mm
  • Bottom plate: 235 × 130 mm