Tambourine
Leverandør: CORNELSEN EXPERIMENTA
46707
765-0157EA 0 NOK
765-0157
Tambourine
Lærings materiell Læremateriale for fysikk Waves and Oscillations Learning Activities
Metal frame with tension spring and membrane of Papyrolin. With the help of two tambourines and a pendulum ball, experiments with pressure waves can be performed.
  • Diameter: 190 mm