Electroscope
Leverandør: CORNELSEN EXPERIMENTA
50090
765-0181EA 0 NOK
765-0181
Electroscope
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
For electrostatic experiments. Consists of insulated aluminum holder with pointed bearing, removable capacitor plate, metal ring with rod and 4 mm earthing contact.
  • Dimension: metal ring diam. 200 mm, rod diam. 10 mm