Plexiglas tube with ball
Leverandør: CORNELSEN EXPERIMENTA
50179
765-0188EA 0 NOK
765-0188
Plexiglas tube with ball
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
Apparatus for demonstrating the movement of a charged body. Consists of plexiglass tubes with two metal electrodes, ball with metallic surface and 4 mm connection sleeves. When connected to a high voltage source, the ball moves back and forth between the electrodes.
  • Length: 150 mm
  • Diam, ball: 43 mm