Thermocouple
Leverandør: CORNELSEN EXPERIMENTA
55085
765-0212EA 0 NOK
765-0212
Thermocouple
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
For experiments with thermoelectric effects. Consists of brazed copper and constant wire. Equipped with connections for 4 mm banana connectors. Mounted on plate with rod, 10 mm.