The charge of the electron, acc. busch
Leverandør: ELWE LEHRGERAETEBAU KLINGENTHA
8481350
765-0597EA 0 NOK
765-0597
The charge of the electron, acc. busch
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
The exercise oscilloscope is placed at the Helmholtz coils so that its main axis coincides with the axis of the coils. A sawtooth-shaped voltage is applied to the deflection plates of the oscilloscope so that a smooth horizontal line appears on the screen. Let direct current pass through the coils at a preset anode voltage and adjust the intensity of the current so that the line is focused at one point. The specific charge e/m of the electron can be calculated from the measured values ​​of the anode voltage and the current through the coil.
  • Determination of the electron's specific