Franck-hertz tries with
Leverandør: ELWE LEHRGERAETEBAU KLINGENTHA
8482150
765-0600EA 0 NOK
765-0600
Franck-hertz tries with
Lærings materiell Læremateriale for fysikk Atomic Physics Learning Activities
Franck-Hertz's experiment is one of the best ways to show the quantum properties of atoms. The experiment verifies the excitation energy of the Hg atoms through collisions with free electrons. Franck-Hertz tubes consist of an oxide cathode, gallery anode and a collection electrode. The evacuated tube contains a drop of Hg. Before starting the measurements, the pipe must be heated to 200 °C, partly to increase the pressure of the Hg steam and partly to obtain a sufficient probability of collision. When the temperature in the tube has become constant, the cathode is heated and a direct voltageMore Product Information