Motor, 12 V DC, with iron-free rotor
Leverandør: ELWE LEHRGERAETEBAU KLINGENTHA
8552330
765-0601EA 0 NOK
765-0601
Motor, 12 V DC, with iron-free rotor
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Motor with rotor that lacks iron and thereby has a very low moment of inertia, which in turn enables fast start and stop or change of direction of rotation. The time from start to maximum speed (3000 rpm) is only 11 ms. Can be used for experiments with mechanical oscillations. The motor is most conveniently operated with a function generator.
  • Rated voltage: 12 V.