Conductor ball
Leverandør: GR NNA PLASTEMBALLAGE AB
192662-0
765-0920EA 0 NOK
765-0920
Conductor ball
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
Very light ball of porous plastic. With thin nylon thread for hanging. Used for influence tests, for transferring charge, eg between capacitor plates etc.
  • Diam: 38 mm
  • Weight: 2 g