Thermistors
Leverandør: GR NNA PLASTEMBALLAGE AB
192897-3
765-0924EA 230 NOK
765-0924
Thermistors
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
Semiconductor type nonlinear resistors with large negative temperature coefficient (NTC resistors). Used for studies of temperature dependence of resistances, such as temperature sensors, current limiters, etc. Of standardized disc type with connecting wires.
  • dR / dT: −4% / ° C (at 25 ° C)
Pmax: 0.5 W
Diam: 5 mm