Experiment Kit, general physics and mechanics
Leverandør: GR NNA PLASTEMBALLAGE AB
194250-42
765-0927EA 0 NOK
765-0927
Experiment Kit, general physics and mechanics
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
The set is primarily intended for the upper level of elementary school or the equivalent level of knowledge. It is suitable for both demonstration and laboratory use. The kit can be used in any room with access to water. The kit includes everything needed with the exception of table salt, rubbing alcohol, cotton and cardboard sheets. The material is stored in a specially designed wooden box. Through a wealth of detail and many combination possibilities, basic physics experiments can be carried out in a number of different areas. Examples of attempts: