Laser with magnetic mount
Leverandør: KVANT LIFE SCIENCE
194570-615
765-1003EA 0 NOK
765-1003
Laser with magnetic mount
Lærings materiell Læremateriale for fysikk Optics Learning Activities
Laser light source for mounting on magnetic board (white board or similar). Can be used to advantage in combination with picture optics (765-1009).
  • Several lasers can be mounted in parallel if a beam is desired