Picture optics, completion kit
Leverandør: KVANT LIFE SCIENCE
21-0411
765-1009EA 0 NOK
765-1009
Picture optics, completion kit
Lærings materiell Læremateriale for fysikk Optics Learning Activities
New optical models for laser optics (15 parts). Consists of.
  • concave lenses
  • plank concave lenses
  • convex lenses
  • planar convex lenses
  • equilateral prism
  • rectangular prisms
  • mirrors
  • condenser lenses
  • cubic beam splitters