Laser communication
Leverandør: KVANT LIFE SCIENCE
21-1001
765-1013EA 0 NOK
765-1013
Laser communication
Lærings materiell Læremateriale for fysikk Waves and Oscillations Learning Activities
Equipment for laser sound transmission experiments. The included microphone, tape recorder or function generator is used as the sound source. The rate consists of.
  • Modular laser (635 nm / 1mW)