Tripod foot, tripod, PASS
Leverandør: PHYWE
02002.55
765-1137EA 0 NOK
765-1137
Tripod foot, tripod, PASS
Stativer
Made of cast, lacquered zinc alloy. Equipped with rubber feet with level control on all three legs. Fits tripod rods with a diameter of 6 to 14 mm.
Dimensions: limitation radius 110 mm, height 50 58 mm
Weight: 1.8 kg