Tripod base, cylindrical, PASS
Leverandør: PHYWE
02006.55
765-1139EA 0 NOK
765-1139
Tripod base, cylindrical, PASS
Stativer
Made of cast iron with coating of reinforced plastic. Intended for cylindrical stand rods with a diameter of 4 to 15 mm or square stand rods with a cross section of 4×4 to 12×12 mm. A 28×9 mm socket in the bottom of the foot is suitable for ordinary wooden scales, which is why the foot can be used to advantage in, for example, optical experiments.
Dimensions: base diameter 70 mm, height 55 mm