Tripod plate, H-foot, PASS
Leverandør: PHYWE
02007.55
765-1140EA 0 NOK
765-1140
Tripod plate, H-foot, PASS
Stativer
Extremely stable H-foot with 5 attachment points and many combination options. Intended for tripod rods 4 to 12 mm or square tripod rods with a cross section of 4×4 mm to 12×12 mm. If a tripod rod is mounted between two standing feet, a compact, stable and easily movable physics bench is obtained.
Dimensions: 193×178×36 mm