Interference tube according to quincke
Leverandør: PHYWE
03482.00
765-1177EA 0 NOK
765-1177
Interference tube according to quincke
Lærings materiell Læremateriale for fysikk Waves and Oscillations Learning Activities
Quincke interference tube for determining wavelengths and frequencies by interfering with sound waves in air. Consists of two U-shaped metal tubes slidable into each other with a measuring scale. Inputs and outputs with knobs. Three brackets for mounting on a stand. Comes with listening hose.
  • Frequency range: 2-5 kHz
  • Dimensions: 930 × 250 mm