Diver, Cartesian
Leverandør: PHYWE
03820.03
765-1178EA 265 NOK
765-1178
Diver, Cartesian
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Divers of glass, placed in a bottle filled with water. If the liquid pressure increases, water flows into the diver, whose average density increases. This results in the diver sinking to rise again towards the surface when the pressure is reduced.