Plate capacitor
Leverandør: PHYWE
06220.00
765-1189EA 67875 NOK
765-1189
Plate capacitor
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
Precision capacitor for electrostatic tests e.g. examination of the relationship between charge, voltage and capacitance. Measurement of the dielectric constant and the electrostatic induction constant. The permanently mounted plate is well insulated while the movable plate is mounted directly in the stand. The distance between the capacitor plates can be adjusted with an accuracy of 0.1 mm. Distance between plates 0 to 70 mm.
  • Plate diameter 260 mm
  • Plate thickness 6 mm