Neon lamps
Leverandør: PHYWE
06656.00
765-1203EA 255 NOK
765-1203
Neon lamps
Lys Neonlampe
Tubular glow lamp of glass. With conical contact hats at the ends. Used, inter alia, to determine the polarity of charge, because light only occurs around the negative electrode. Ignition voltage approximately 80 V, switching voltage approximately 65 V. For example, connects with crocodile clips.
Length: 50 mm
Ø: 10 mm