Thermos stack according to Moll
Leverandør: PHYWE
08479.00
765-1228EA 0 NOK
765-1228
Thermos stack according to Moll
Lærings materiell Læremateriale for fysikk Heat/Thermodynamics Learning Activities
Used to detect thermal radiation, visible light and ultraviolet light. The sensitivity is wavelength dependent in the range 150 nm-15 µm. The output voltage is closely proportional to the incident radiant energy. Suitable for measuring small temperature differences. Quick stabilization, 2-3 p. Consists of 16 series-connected thermocouples placed in a conical reflector. The elements are protected by a high-gloss metal cylinder, which is fitted with a removable protective glass at the front. Suitable for use with, for example, microvolt amplifiers 765-1293 in conjunction with demonstrationMore Product Information