Microvolt amplifier for demonstration instruments
Leverandør: PHYWE
11110.01
765-1293EA 0 NOK
765-1293
Microvolt amplifier for demonstration instruments
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
For measuring low currents AC/DC and DC voltages AC/DC eg Hall voltages and thermal voltages. High sensitivity and very good electrical stability. Well shielded from disturbing voltages. Overload protected up to 220 V AC/DC. The front is equipped with two scales 0 to 3 and 0 to 10 both with fine division, measuring area window and automatic function indication. The back is equipped with a function switch, measuring range switch, pushbutton for zero setting and steering wheel for electrical zero adjustment. There is also a printer connection.
Measuring ranges: 0 to 30 µV
0 to 100 µV