Voltmeter, electrostatic
Leverandør: PHYWE
11150.00 11151.00
765-1296EA 0 NOK
765-1296 765-1297
Voltmeter, electrostatic
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
Demonstration instrument for lossless measurement of DC and AC voltages. Due to the high insulation resistance (approx. 1014 ©), the instrument loads the voltage source insignificantly. At the same time, the voltage drop becomes negligible. The measuring systems are protected against disturbing fields and placed in plastic apparatus housings. The measurement can be studied from the back through a window. The large and clear scale and the design of the pointer make the instrument very easy to read even at longer distances. The scaling of the scale enables precision reading. Scale is alsoMore Product Information