Arrow board with falling target
Leverandør: PHYWE
11218.00
765-1304EA 0 NOK
765-1304
Arrow board with falling target
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Apparatus for demonstrating the independence of the components of a composite motion. A force is exerted against an arrow, in the direction of a vertically set target, by means of a wire connected to an elastic band. When the arrow is released, the target is released at the same time. The arrow always hits the free-falling target, regardless of the position from which the arrow is released. Supplied with wooden plate with mounted strap, spring lock for attaching the target, arrow with string (approx. 275 cm) and target.
  • Width: 150 mm
  • Height: 860 mm