Ballistic pendulum and throwing device
Leverandør: PHYWE
11229.00
765-1305EA 29725 NOK
765-1305
Ballistic pendulum and throwing device
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
With a few simple steps, the ballistic pendulum is quickly rebuilt into a throwing device with a variable/variable firing angle. The ballistic pendulum, a common method for determining the speed of a pistol bullet, is to push them into a sandbag. With the ballistic pendulum, a harmless variant of this experiment can be performed in the classroom.
  • A steel ball is pushed horizontally against a pendulum and is caught by it. The maximum angular angle of the pendulum is then read on a special scale
  • The projectile mass, pendulum mass and maximum angular range