Table optics, TESS OE1
Leverandør: PHYWE
13276.88
765-1317EA 0 NOK
765-1317
Table optics, TESS OE1
Lærings materiell Læremateriale for fysikk Optics Learning Activities
PHYWE table optics for laboratory work with mirrors, light refraction, lenses, eye function and light refraction. Content:
  • 1 light box with halogen lamp 12 V/20 W
  • 2 columns
  • 6 lens models
  • 1 cuvette, double, semicircular
  • 1 mirror, plan
  • 1 mirror, concave-convex
  • 1 optical disc
  • 1 storage box
  • Delivered in plywood box