Zeeman effects
Leverandør: PHYWE
99999
765-1333EA 0 NOK
765-1333
Zeeman effects
Lærings materiell Læremateriale for fysikk Optics Learning Activities
With equipment, demonstration and accurate calculations can be performed of how the atom's spectral lines are divided by the influence of a magnetic field (the Zeeman effect).
  • The simplest variant is the division of a spectral line into three components (the 'normal Zeeman effect'). In this experiment a Cd spectral lamp with 1 = 634.8 nm is used. The CD lamp is subjected to varying magnetic flux density and the division of the CD line is examined with a Fabry-Perot interferrometer. This wavelength corresponds to a D-P transition (singular transition).