Gas chromatograph, GC-82-12
Leverandør: EDUCATIONSCIENCE COLLECTION
ML30821200
766-0138EA 0 NOK
766-0138
Gas chromatograph, GC-82-12
Kromatografisystemer Gasskromatografer
GC82-12 Hot wire. A simple gas chromatograph with hot wire detector. Compact and easy to handle and suitable for teaching situations. Helium is recommended as the carrier gas, but nitrogen or argon can also be used. Digital temperature setting of the isothermal oven. Temperature control with PT100 sensor 0 to 250 ° C. Adjustable hot wire detector 50 to 300 mA. Two-column system that is also suitable for glass columns. 2 packed copper columns, polar and non-polar, included. Inlet gas inlet pressure 1.5 to 3.5 bar. Dimensions: 30 × 54 × 36 cm. Weight: 11 kg.
Feel free to contact us if youMore Product Information