Saltsyre 6 mol/l (6 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Warning

7710.9010EA 0 NOK
7710.9010
Saltsyre 6 mol/l (6 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Saltsyre
Formel: HCl
Tetthet: 1,06 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance of solution clear, colourless
Normality (N) 5.9 - 6.1

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR