Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular, GenAR® USP, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Warning

Synonymer: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

7730-06EA 3000 NOK
7730-06 7730-88
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular, GenAR® USP, Macron Fine Chemicals™
D(+)-Glukose
Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Smeltepunkt: 146
Tetthet: 1.54 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00148912
CAS-nummer: 50-99-7
EINECS: 200-075-1
Merck Indeks: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C: Water (H₂O) ≤1.0 %
USP - Assay (Anhydrous basis) 97.5 - 102.0 %
USP - Related substances: Maltose and isomaltose ≤0.4 %
USP - Dextrin Passes Test
USP - Related substances: Maltotriose ≤0.2 %
USP - Related substances: Fructose ≤0.15 %
USP - Related substances: Unspecified impurities, each ≤0.10 %
USP - Related substances: Total Impurities ≤0.5 %
USP - Color and Clarity of Solution Passes Test
USP - Soluble Starch, Sulfites ≤15 ppm
USP - Conductivity (uS cm-1) ≤20 µS/cm
EP/BP - Identification A (Specific Rotation (+)) 52.5 - 53.3 Degree
EP/BP - Identification B Passes Test
EP/BP - Identification E Passes Test
EP/BP - Assay (C₆H₁₂O₆) (anhydrous substance) 97.5 - 102.0 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Conductivity, uS cm-1 ≤20
EP/BP - Related Substances: sum of impurities A and B ≤0.4 %
EP/BP - Related Substances: impurity C ≤0.2 %
EP/BP - Related Substances: impurity D ≤0.15 %
EP/BP - Related Substances: unspecified impurities, each ≤0.10 %
EP/BP - Related Substances: total impurities ≤0.5 %
EP/BP - Dextrins Passes Test
EP/BP - Soluble starch, sulfite ≤15 ppm
EP/BP - Water (H₂O) ≤1.0 %
Endotoxin Concentration (EU/g) ≤5.0
JP - Identification 1 Passes Test
JP - Identification 2 Passes Test
JP - Water (H₂O) ≤1.0 %
JP - Assay (Anhydrous basis) 97.5 - 102.0 %
JP - Maltose and isomaltose ≤0.4 %
JP - Maltotriose ≤0.2 %
JP - Fructose ≤0.15 %
JP - Unspecified Impurities, each ≤0.10 %
JP - Total Impurities ≤0.5 %
JP - Color and Clarity of Solution Passes Test
JP - Soluble Starch, Sulfites ≤15 ppm
JP - Conductivity (uS cm-1) ≤20 µS/cm
JP - Dextrin Passes Test
JP - Heavy Metals (as Pb) ≤4 ppm
Appearance (white, granular powder)
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR